siRNA Details
| Gene Symbol: Mapk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031622.2 | NP_113810.2 | 2 | 988 | ||
| XM_006243409.2 | XP_006243471.1 | 2 | 1482 | |||
| GenBank mRNA | BC109380.1 | - | 2 | 939 | 7446401 | |
| M64301.1 | - | 1 | 616 | |||

Mapk6
Genomic Map