siRNA Details
| Gene Symbol: Mapk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031622.2 | NP_113810.2 | 2, 3 | 1324 | ||
| XM_006243409.2 | XP_006243471.1 | 2, 3 | 1818 | |||
| GenBank mRNA | BC109380.1 | - | 2, 3 | 1275 | 7446401 | |
| M64301.1 | - | 1, 2 | 952 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_015806, NM_027418, BC100385, BC006778, X64607, and others.

Mapk6
Genomic Map