siRNA Details
| Gene Symbol: Grb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031623.1 | NP_113811.1 | 3 | 476 | ||
| GenBank mRNA | AF076619.1 | - | 3 | 476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grb14| Gene Symbol: Grb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031623.1 | NP_113811.1 | 3 | 476 | ||
| GenBank mRNA | AF076619.1 | - | 3 | 476 | ||