siRNA Details
| Gene Symbol: Mefv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031634.1 | NP_113822.1 | 2 | 433 | ||
| GenBank mRNA | AF143410.1 | - | 2 | 433 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mefv| Gene Symbol: Mefv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031634.1 | NP_113822.1 | 2 | 433 | ||
| GenBank mRNA | AF143410.1 | - | 2 | 433 | ||