siRNA Details
| Gene Symbol: Fut2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031635.1 | NP_113823.1 | 3 | 344 | ||
| XM_006229130.2 | XP_006229192.1 | 2 | 240 | |||
| XM_006229135.2 | XP_006229197.1 | 3 | 332 | |||
| XM_006229136.2 | XP_006229198.1 | 2 | 243 | |||
| XM_006229137.2 | XP_006229199.1 | 2 | 3092 | |||
| XM_006229138.2 | XP_006229200.1 | 2 | 1498 | |||
| XM_006229139.2 | XP_006229201.1 | 2 | 3353 | |||
| XM_006229140.2 | XP_006229202.1 | 2 | 219 | |||
| XM_006229141.2 | XP_006229203.1 | 2 | 1488 | |||
| XM_008759412.1 | XP_008757634.1 | 2 | 351 | |||
| GenBank mRNA | AB006138.1 | - | 3 | 344 | ||
| AF042743.1 | - | 1 | 50 | |||
| AF264005.1 | - | 1 | 131 | |||

Fut2
Genomic Map