siRNA Details
| Gene Symbol: Fut2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031635.1 | NP_113823.1 | 3 | 1392 | ||
| XM_006229130.2 | XP_006229192.1 | 2 | 1288 | |||
| XM_006229135.2 | XP_006229197.1 | 3 | 1380 | |||
| XM_006229136.2 | XP_006229198.1 | 2 | 1291 | |||
| XM_006229137.2 | XP_006229199.1 | 2 | 4140 | |||
| XM_006229138.2 | XP_006229200.1 | 2 | 2546 | |||
| XM_006229139.2 | XP_006229201.1 | 2 | 4401 | |||
| XM_006229140.2 | XP_006229202.1 | 2 | 1267 | |||
| XM_006229141.2 | XP_006229203.1 | 2 | 2536 | |||
| XM_008759412.1 | XP_008757634.1 | 2 | 1399 | |||
| GenBank mRNA | AB006138.1 | - | 3 | 1392 | ||
| L26010.1 | - | 1 | 519 | |||

Fut2
Genomic Map