siRNA Details
| Gene Symbol: Hpgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031644.2 | NP_113832.1 | 2, 3 | 499 | ||
| XM_006236602.2 | XP_006236664.1 | 2, 3 | 467 | |||
| GenBank mRNA | AF021882.1 | - | 1, 2 | 27 | ||
| BC087590.1 | - | 2, 3 | 499 | 7307804 | ||
| D82071.1 | - | 2, 3 | 330 | |||

Hpgds
Genomic Map