siRNA Details
Gene Symbol: Hpgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031644.2 | NP_113832.1 | 6 | 1153 | ||
XM_006236602.2 | XP_006236664.1 | 6 | 1124 | |||
XM_006236603.2 | XP_006236665.1 | 5 | 1046 | |||
GenBank mRNA | BC087590.1 | - | 6 | 1153 | 7307804 | |
D82071.1 | - | 6 | 984 |
