siRNA Details
| Gene Symbol: Ramp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031645.1 | NP_113833.1 | 2 | 134 | ||
| XM_006245473.2 | XP_006245535.1 | 2 | 998 | |||
| XM_006245474.2 | XP_006245536.1 | 2 | 210 | |||
| GenBank mRNA | AB030942.1 | - | 2 | 110 | ||
| AB042887.1 | - | 2 | 134 | |||
| AF181550.1 | - | 2 | 111 | |||

Ramp1
Genomic Map