siRNA Details
| Gene Symbol: Ramp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031646.1 | NP_113834.1 | 3 | 286 | ||
| GenBank mRNA | AB030943.1 | - | 3 | 295 | ||
| AB042888.1 | - | 3 | 286 | |||
| AF162778.1 | - | 1 | 40 | |||
| AF181551.1 | - | 3 | 289 | |||
| EF595744.1 | - | 3 | 210 | |||
| FQ226087.1 | - | 3 | 311 | |||

Ramp2
Genomic Map