siRNA Details
| Gene Symbol: Fxyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031648.2 | NP_113836.2 | 3 | 170 | ||
| XM_006228838.2 | XP_006228900.1 | 4 | 390 | |||
| XM_006228839.2 | XP_006228901.1 | 3 | 165 | |||
| XM_008759184.1 | XP_008757406.1 | 3 | 179 | |||
| GenBank mRNA | CB712570.1 | - | 3 | 170 | ||
| FM109156.1 | - | 3 | 124 | |||
| FQ211349.1 | - | 3 | 181 | |||
| FQ224131.1 | - | 3 | 133 | |||
| FQ224338.1 | - | 3 | 216 | |||
| U72246.1 | - | 2 | 84 | |||

Fxyd1
Genomic Map