siRNA Details
| Gene Symbol: Klrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031649.2 | NP_113837.1 | 1, 2 | 90 | ||
| GenBank mRNA | X79812.1 | - | 1, 2 | 143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klrg1| Gene Symbol: Klrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031649.2 | NP_113837.1 | 1, 2 | 90 | ||
| GenBank mRNA | X79812.1 | - | 1, 2 | 143 | ||