siRNA Details
| Gene Symbol: Kcnab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031652.1 | NP_113840.1 | 4 | 752 | ||
| XM_008767862.1 | XP_008766084.1 | 4 | 790 | |||
| GenBank mRNA | X76723.1 | - | 4 | 752 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnab3| Gene Symbol: Kcnab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031652.1 | NP_113840.1 | 4 | 752 | ||
| XM_008767862.1 | XP_008766084.1 | 4 | 790 | |||
| GenBank mRNA | X76723.1 | - | 4 | 752 | ||