siRNA Details
| Gene Symbol: Grk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112712.1 | NP_001106183.1 | 6 | 507 | ||
| NM_001112713.1 | NP_001106184.1 | 6 | 507 | |||
| NM_031657.3 | NP_113845.2 | 6 | 507 | |||
| XM_006253653.2 | XP_006253715.1 | 6 | 616 | |||
| XM_008771504.1 | XP_008769726.1 | 5 | 774 | |||
| GenBank mRNA | AF040750.1 | - | 6 | 507 | ||
| EX488847.1 | - | 6 | 667 | |||
| X97439.1 | - | 6 | 507 | |||
| X97440.1 | - | 6 | 507 | |||
| Y09365.1 | - | 6 | 535 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001038018, NM_001286064, NM_001286063, NM_001286066, NM_001112711, and others.

Grk6
Genomic Map