siRNA Details
Gene Symbol: Kcnk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033376.1 | NP_203694.1 | 2 | 1581 | ||
GenBank mRNA | AF031384.1 | - | 2 | 1581 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033376.1 | NP_203694.1 | 2 | 1581 | ||
GenBank mRNA | AF031384.1 | - | 2 | 1581 |