siRNA Details
| Gene Symbol: Igf1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052807.2 | NP_434694.1 | 21 | 4258 | ||
| XM_006229324.2 | XP_006229386.1 | 21 | 4261 | |||
| XM_008759500.1 | XP_008757722.1 | 21 | 4262 | |||
| GenBank mRNA | L29232.1 | - | 21 | 4257 | ||

Igf1r
Genomic Map