siRNA Details
| Gene Symbol: TRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012288.3 | NP_036420.1 | 5 | 572 | ||
| XM_011515005.2 | XP_011513307.1 | 5 | 447 | |||
| GenBank mRNA | AB621814.1 | - | 5 | 568 | ||
| AK291751.1 | - | 5 | 458 | |||
| AK311560.1 | - | 5 | 441 | |||
| BC028121.1 | - | 5 | 572 | 5169808 | ||
| D31762.1 | - | 5 | 498 | |||
| KU178488.1 | - | 5 | 423 | |||
| KU178489.1 | - | 4 | 359 | |||

TRAM2
Genomic Map