siRNA Details
| Gene Symbol: Glrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053296.1 | NP_445748.1 | 6 | 628 | ||
| XM_006232506.2 | XP_006232568.1 | 7 | 774 | |||
| GenBank mRNA | AJ310839.1 | - | 6 | 628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glrb| Gene Symbol: Glrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053296.1 | NP_445748.1 | 6 | 628 | ||
| XM_006232506.2 | XP_006232568.1 | 7 | 774 | |||
| GenBank mRNA | AJ310839.1 | - | 6 | 628 | ||