siRNA Details
| Gene Symbol: Pkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053297.2 | NP_445749.1 | 6, 7 | 926 | ||
| XM_006243187.2 | XP_006243249.1 | 6, 7 | 1156 | |||
| XM_006243188.2 | XP_006243250.1 | 6, 7 | 1156 | |||
| XM_006243190.2 | XP_006243252.1 | 7, 8 | 1228 | |||
| GenBank mRNA | AY724474.1 | - | 6, 7 | 923 | ||
| FQ223721.1 | - | 6, 7 | 934 | |||
| FQ224316.1 | - | 3, 4 | 584 | |||
| X15800.1 | - | 6, 7 | 928 | |||
| Gene Symbol: LOC100362738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772937.1 | XP_008771159.1 | 1 | 890 | ||
| Gene Symbol: LOC100364062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751952.3 | XP_003752000.1 | 1 | 850 | ||

Pkm
Genomic Map