siRNA Details
| Gene Symbol: Gpr182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053302.3 | NP_445754.2 | 2 | 262 | ||
| XM_008765379.1 | XP_008763601.1 | 2 | 576 | |||
| XM_008765380.1 | XP_008763602.1 | 1 | 626 | |||
| GenBank mRNA | BC129072.1 | - | 2 | 262 | 7096756 | |
| S79811.1 | - | 2 | 340 | |||
More Information
Set Membership: |
|

Gpr182
Genomic Map