siRNA Details
| Gene Symbol: Hpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053318.1 | NP_445770.1 | 2 | 153 | ||
| GenBank mRNA | BC091137.1 | - | 2 | 144 | 7326169 | |
| FQ210231.1 | - | 2 | 154 | |||
| FQ211176.1 | - | 2 | 154 | |||
| FQ218633.1 | - | 2 | 152 | |||
| FQ218971.1 | - | 2 | 146 | |||
| FQ219309.1 | - | 2 | 152 | |||

Hpx
Genomic Map