siRNA Details
| Gene Symbol: Cd247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205304.1 | NP_001192233.1 | 7 | 992 | ||
| NM_170789.2 | NP_740770.2 | 8 | 1073 | |||
| XM_006250147.2 | XP_006250209.1 | 8 | 1113 | |||
| GenBank mRNA | BC097933.1 | - | 8 | 1016 | 7384006 | |
| D13555.1 | - | 8 | 1074 | |||

Cd247
Genomic Map