siRNA Details
Gene Symbol: Canx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172008.2 | NP_742005.1 | 11 | 1309 | ||
GenBank mRNA | L18889.1 | - | 10 | 1206 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
