siRNA Details
| Gene Symbol: Myo1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173101.1 | NP_775124.1 | 2 | 360 | ||
| GenBank mRNA | X74815.1 | - | 2 | 360 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo1e| Gene Symbol: Myo1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173101.1 | NP_775124.1 | 2 | 360 | ||
| GenBank mRNA | X74815.1 | - | 2 | 360 | ||