siRNA Details
| Gene Symbol: Cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175577.3 | NP_783167.2 | 7 | 1256 | ||
| XM_006231021.2 | XP_006231083.1 | 7 | 1363 | |||
| XM_008760218.1 | XP_008758440.1 | 7 | 1363 | |||
| XM_008760219.1 | XP_008758441.1 | 6 | 1059 | |||
| GenBank mRNA | AF488727.1 | - | 7 | 1195 | ||
| AY683561.1 | - | 6 | 875 | |||
| BC089767.1 | - | 7 | 1256 | 7383438 | ||

Cd6
Genomic Map