siRNA Details
| Gene Symbol: Cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175577.3 | NP_783167.2 | 13 | 2638 | ||
| XM_006231021.2 | XP_006231083.1 | 13 | 2742 | |||
| XM_008760218.1 | XP_008758440.1 | 12 | 2628 | |||
| XM_008760219.1 | XP_008758441.1 | 12 | 2441 | |||
| GenBank mRNA | AF488727.1 | - | 13 | 2624 | ||
| BC089767.1 | - | 10 | 3142 | 7383438 | ||

Cd6
Genomic Map