siRNA Details
| Gene Symbol: Atp1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012507.3 | NP_036639.2 | 3 | 850 | ||
| XM_006246580.2 | XP_006246642.1 | 3 | 873 | |||
| XM_006246581.2 | XP_006246643.1 | 3 | 873 | |||
| GenBank mRNA | BC087034.1 | - | 3 | 850 | 7124310 | |
| J04629.1 | - | 3 | 739 | |||
| U45946.1 | - | 3 | 288 | |||

Atp1b2
Genomic Map