siRNA Details
| Gene Symbol: C4bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012516.2 | NP_036648.2 | 8 | 1111 | ||
| GenBank mRNA | BC088165.1 | - | 8 | 1111 | 7324823 | |
| Z50051.1 | - | 6 | 774 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C4bpa| Gene Symbol: C4bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012516.2 | NP_036648.2 | 8 | 1111 | ||
| GenBank mRNA | BC088165.1 | - | 8 | 1111 | 7324823 | |
| Z50051.1 | - | 6 | 774 | |||