siRNA Details
| Gene Symbol: Camk2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012519.2 | NP_036651.1 | 9 | 887 | ||
| XM_006233284.2 | XP_006233346.1 | 9 | 1106 | |||
| XM_006233285.2 | XP_006233347.1 | 9 | 1106 | |||
| XM_008761452.1 | XP_008759674.1 | 9 | 1106 | |||
| GenBank mRNA | BC107562.1 | - | 749 | 7939424 | ||
| J05072.1 | - | 887 | ||||

Camk2d
Genomic Map