siRNA Details
| Gene Symbol: Cd53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012523.2 | NP_036655.1 | 7 | 680 | ||
| GenBank mRNA | BC078900.1 | - | 7 | 631 | 7130719 | |
| M57276.1 | - | 7 | 694 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd53| Gene Symbol: Cd53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012523.2 | NP_036655.1 | 7 | 680 | ||
| GenBank mRNA | BC078900.1 | - | 7 | 631 | 7130719 | |
| M57276.1 | - | 7 | 694 | |||