siRNA Details
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012529.3 | NP_036661.3 | 325 | |||
| GenBank mRNA | BC065307.1 | - | 307 | 5598839 | ||
| BC070955.1 | - | 312 | 7104205 | |||
| BC087656.1 | - | 324 | ||||
| BC099814.1 | - | 309 | 7929525 | |||
| BC127477.1 | - | 315 | 8367180 | |||
| FM060701.1 | - | 330 | ||||
| FQ213834.1 | - | 300 | ||||
| FQ216091.1 | - | 1 | 44 | |||
| M14400.1 | - | 337 | ||||
| M57664.1 | - | 2 | 188 | |||

Ckb
Genomic Map