siRNA Details
| Gene Symbol: Comt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012531.2 | NP_036663.1 | 2 | 298 | ||
| XM_006248603.2 | XP_006248665.1 | 2 | 299 | |||
| GenBank mRNA | BC081850.1 | - | 2 | 156 | 7191956 | |
| CK478774.1 | - | 2 | 418 | |||
| FQ209401.1 | - | 11 | ||||
| FQ209638.1 | - | 11 | ||||
| FQ209670.1 | - | 11 | ||||
| FQ209706.1 | - | 1 | 22 | |||
| FQ210416.1 | - | 1 | 47 | |||
| FQ210430.1 | - | 1 | 14 | |||
| FQ218190.1 | - | 1 | 13 | |||
| FQ218375.1 | - | 1 | 21 | |||
| FQ218564.1 | - | 1 | 13 | |||
| FQ218595.1 | - | 1 | 13 | |||
| FQ219155.1 | - | 1 | 18 | |||
| FQ219257.1 | - | 1 | 18 | |||
| FQ219433.1 | - | 1 | 13 | |||
| FQ223536.1 | - | 1 | 13 | |||
| FQ228829.1 | - | 11 | ||||
| FQ228870.1 | - | 1 | 19 | |||
| M60753.1 | - | 102 | ||||

Comt
Genomic Map