siRNA Details
| Gene Symbol: Cyp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012540.2 | NP_036672.2 | 2 | 590 | ||
| XM_006243150.2 | XP_006243212.1 | 2 | 587 | |||
| GenBank mRNA | X00469.1 | - | 2 | 590 | ||
| X17160.1 | - | 600 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp1a1| Gene Symbol: Cyp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012540.2 | NP_036672.2 | 2 | 590 | ||
| XM_006243150.2 | XP_006243212.1 | 2 | 587 | |||
| GenBank mRNA | X00469.1 | - | 2 | 590 | ||
| X17160.1 | - | 600 | ||||