siRNA Details
| Gene Symbol: Cyp2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012542.2 | NP_036674.1 | 1 | 152 | ||
| GenBank mRNA | J02852.1 | - | 1 | 131 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2a3| Gene Symbol: Cyp2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012542.2 | NP_036674.1 | 1 | 152 | ||
| GenBank mRNA | J02852.1 | - | 1 | 131 | ||