siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012557.2 | NP_036689.2 | 4, 5 | 600 | ||
| XM_006253488.2 | XP_006253550.1 | 5, 6 | 875 | |||
| XM_006253489.2 | XP_006253551.1 | 4, 5 | 732 | |||
| XM_006253490.2 | XP_006253552.1 | 4, 5 | 577 | |||
| XM_006253491.2 | XP_006253553.1 | 1, 2 | 1406 | |||
| XM_008771423.1 | XP_008769645.1 | 3, 4 | 1722 | |||
| GenBank mRNA | BC168147.1 | - | 4, 5 | 600 | 9024689 | |
| U73586.1 | - | 3, 4 | 376 | |||

Fancc
Genomic Map