siRNA Details
| Gene Symbol: Glra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012568.2 | NP_036700.1 | 3, 4 | 825 | ||
| XM_003752039.3 | XP_003752087.1 | 4, 5 | 449 | |||
| XM_008773153.1 | XP_008771375.1 | 3, 4 | 231 | |||
| GenBank mRNA | AJ310837.1 | - | 3, 4 | 274 | ||
| X61159.1 | - | 3, 4 | 825 | |||

Glra2
Genomic Map