siRNA Details
| Gene Symbol: Got1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | J04171.1 | - | 3 | 383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Got1| Gene Symbol: Got1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | J04171.1 | - | 3 | 383 | ||