siRNA Details
| Gene Symbol: Grin2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012573.3 | NP_036705.3 | 3 | 689 | ||
| GenBank mRNA | D13211.1 | - | 2 | 482 | ||
| M91561.1 | - | 3 | 689 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grin2a| Gene Symbol: Grin2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012573.3 | NP_036705.3 | 3 | 689 | ||
| GenBank mRNA | D13211.1 | - | 2 | 482 | ||
| M91561.1 | - | 3 | 689 | |||