siRNA Details
Gene Symbol: Grin2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012574.1 | NP_036706.1 | 3 | 1071 | ||
GenBank mRNA | M91562.1 | - | 3 | 778 | ||
U11419.1 | - | 3 | 1071 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Grin2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012574.1 | NP_036706.1 | 3 | 1071 | ||
GenBank mRNA | M91562.1 | - | 3 | 778 | ||
U11419.1 | - | 3 | 1071 |