siRNA Details
| Gene Symbol: Iapp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237614.2 | XP_006237676.1 | 3 | 1192 | ||
| GenBank mRNA | J04544.1 | - | 3 | 750 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iapp| Gene Symbol: Iapp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237614.2 | XP_006237676.1 | 3 | 1192 | ||
| GenBank mRNA | J04544.1 | - | 3 | 750 | ||