siRNA Details
| Gene Symbol: Ldhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012595.1 | NP_036727.1 | 7 | 765 | ||
| XM_006237615.2 | XP_006237677.1 | 8 | 936 | |||
| GenBank mRNA | BC059149.1 | - | 7 | 822 | 6919339 | |
| FQ212899.1 | - | 7 | 828 | |||
| FQ214847.1 | - | 7 | 812 | |||
| FQ215687.1 | - | 7 | 828 | |||
| U07181.1 | - | 7 | 765 | |||

Ldhb
Genomic Map