siRNA Details
| Gene Symbol: Lipc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012597.2 | NP_036729.2 | 3 | 488 | ||
| XM_006243359.2 | XP_006243421.1 | 5 | 591 | |||
| XM_006243360.2 | XP_006243422.1 | 4 | 543 | |||
| GenBank mRNA | BC088160.1 | - | 3 | 488 | 7323251 | |
| M16235.1 | - | 3 | 410 | |||
| X17366.1 | - | 3 | 398 | |||

Lipc
Genomic Map