siRNA Details
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012598.2 | NP_036730.1 | 2 | 415 | ||
GenBank mRNA | BC081836.1 | - | 2 | 415 | 7106121 | |
L03294.1 | - | 400 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012598.2 | NP_036730.1 | 2 | 415 | ||
GenBank mRNA | BC081836.1 | - | 2 | 415 | 7106121 | |
L03294.1 | - | 400 |