siRNA Details
| Gene Symbol: Npr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012613.1 | NP_036745.1 | 2 | 1164 | ||
| XM_006232591.2 | XP_006232653.1 | 3 | 1665 | |||
| GenBank mRNA | BC128742.1 | - | 2 | 1150 | 7458324 | |
| M74535.1 | - | 3 | 1113 | |||
| X14773.1 | - | 2 | 1021 | |||

Npr1
Genomic Map