siRNA Details
| Gene Symbol: Npr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012613.1 | NP_036745.1 | 2, 3 | 1287 | ||
| XM_006232591.2 | XP_006232653.1 | 3, 4 | 1788 | |||
| GenBank mRNA | BC128742.1 | - | 2, 3 | 1273 | 7458324 | |
| M74535.1 | - | 3, 4 | 1236 | |||
| X14773.1 | - | 2, 3 | 1144 | |||

Npr1
Genomic Map