siRNA Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012619.2 | NP_036751.2 | 10 | 1106 | ||
| XM_008765215.1 | XP_008763437.1 | 8 | 1046 | |||
| GenBank mRNA | BC078881.1 | - | 10 | 1106 | 7131414 | |
| FQ219397.1 | - | 6 | 542 | |||
| FQ219680.1 | - | 10 | 1138 | |||
| K02599.1 | - | 5 | 396 | |||
| M12337.1 | - | 10 | 1091 | |||

Pah
Genomic Map