siRNA Details
| Gene Symbol: Prkcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012628.1 | NP_036760.1 | 4 | 526 | ||
| XM_006228014.2 | XP_006228076.1 | 4 | 594 | |||
| GenBank mRNA | BC089226.1 | - | 4 | 560 | 7319243 | |
| M13707.1 | - | 4 | 526 | |||
| X07287.1 | - | 4 | 449 | |||

Prkcg
Genomic Map