siRNA Details
| Gene Symbol: BLOC1S6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001311255.1 | NP_001298184.1 | 2 | 283 | ||
| NM_001311256.1 | NP_001298185.1 | 2 | 283 | |||
| NM_012388.3 | NP_036520.1 | 2 | 449 | |||
| NR_132350.1 | 2 | 283 | ||||
| NR_132351.1 | 2 | 449 | ||||
| NR_132352.1 | 2 | 449 | ||||
| NR_132356.1 | 2 | 151 | ||||
| NR_132357.1 | 2 | 151 | ||||
| GenBank mRNA | AF080470.1 | - | 2 | 353 | ||
| AK057545.1 | - | 2 | 193 | |||
| AK128626.1 | - | 2 | 252 | |||
| AK315581.1 | - | 2 | 167 | |||
| AL596952.1 | - | 2 | 222 | |||
| BC004819.1 | - | 2 | 170 | 3450397 | ||
| DA247079.1 | - | 2 | 169 | |||
| DA468279.1 | - | 2 | 449 | |||
| DA523523.1 | - | 2 | 97 | |||
| DA859617.1 | - | 2 | 98 | |||
| DB289156.1 | - | 2 | 94 | |||

BLOC1S6
Genomic Map