siRNA Details
| Gene Symbol: PFDN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012394.3 | NP_036526.2 | 3 | 244 | ||
| XM_011509624.2 | XP_011507926.1 | 4 | 257 | |||
| GenBank mRNA | AF117237.1 | - | 3 | 200 | ||
| AF151065.1 | - | 3 | 184 | |||
| AF165883.1 | - | 3 | 217 | |||
| AK315736.1 | - | 3 | 218 | |||
| BC012464.1 | - | 3 | 213 | 4538003 | ||
| BC047042.1 | - | 3 | 210 | 5541226 | ||
| BI597083.1 | - | 3 | 277 | |||

PFDN2
Genomic Map