siRNA Details
| Gene Symbol: Pthlh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012636.1 | NP_036768.1 | 412 | |||
| GenBank mRNA | M21967.1 | - | 338 | |||
| M31603.1 | - | 412 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pthlh| Gene Symbol: Pthlh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012636.1 | NP_036768.1 | 412 | |||
| GenBank mRNA | M21967.1 | - | 338 | |||
| M31603.1 | - | 412 | ||||